Assay Details
| Gene Symbol: GCGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120592.2 | XP_014976078.2 | 594 | 85 | ||
| XM_015120593.2 | XP_014976079.2 | 459 | 85 | |||
| XM_028837000.1 | XP_028692833.1 | 459 | 85 | |||
| XM_028837001.1 | XP_028692834.1 | 459 | 85 | |||
More Information
Set Membership: |
|

GCGR
Genomic Map