Assay Details
| Gene Symbol: GCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092919.3 | XP_001092919.1 | 377 | 69 | ||
| XM_001093035.4 | XP_001093035.2 | 2218 | 69 | |||
| XM_028845839.1 | XP_028701672.1 | 377 | 69 | |||
| XM_028845840.1 | XP_028701673.1 | 377 | 69 | |||
More Information
Set Membership: |
|

GCK
Genomic Map