Assay Details
| Gene Symbol: LOC100428939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143408.2 | XP_014998894.1 | 4018 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100428939| Gene Symbol: LOC100428939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143408.2 | XP_014998894.1 | 4018 | 115 | ||
Set Membership: |
|