Assay Details
| Gene Symbol: PTEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260965.1 | NP_001247894.1 | 1221 | 73 | ||
| XM_015147613.2 | XP_015003099.1 | 886 | 73 | |||
| XM_028853389.1 | XP_028709222.1 | 1117 | 73 | |||
| XM_028853390.1 | XP_028709223.1 | 557 | 73 | |||
| GenBank mRNA | JU335605.1 | - | 1522 | 73 | ||
| JU471276.1 | - | 1936 | 73 | |||
| JV047745.1 | - | 1221 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTEN
Genomic Map