Assay Details
| Gene Symbol: TNNI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085820.4 | XP_001085820.1 | 256 | 69 | ||
| XM_028838982.1 | XP_028694815.1 | 259 | 69 | |||
| GenBank mRNA | JV703948.1 | - | 231 | 69 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TNNI3
Genomic Map