Assay Details
| Gene Symbol: WAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127369.2 | XP_014982855.1 | 565 | 109 | ||
| XM_015127370.2 | XP_014982856.1 | 568 | 109 | |||
| GenBank mRNA | JU477087.1 | - | 543 | 109 | ||
| JU492849.1 | - | 543 | 109 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

WAS
Genomic Map