Assay Details
| Gene Symbol: KRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098486.3 | XP_001098486.2 | 904 | 112 | ||
| GenBank mRNA | JU475529.1 | - | 847 | 112 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT2| Gene Symbol: KRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098486.3 | XP_001098486.2 | 904 | 112 | ||
| GenBank mRNA | JU475529.1 | - | 847 | 112 | ||
Set Membership: |
|