Assay Details
| Gene Symbol: KRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151781.2 | XP_015007267.2 | 1831 | 135 | ||
| GenBank mRNA | JU486232.1 | - | 1525 | 135 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT5| Gene Symbol: KRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151781.2 | XP_015007267.2 | 1831 | 135 | ||
| GenBank mRNA | JU486232.1 | - | 1525 | 135 | ||
Set Membership: |
|