Assay Details
| Gene Symbol: NEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266298.1 | NP_001253227.1 | 1107 | 113 | ||
| XM_028846845.1 | XP_028702678.1 | 816 | 113 | |||
| GenBank mRNA | JU334658.1 | - | 1107 | 113 | ||
| JU475713.1 | - | 1118 | 113 | |||
| JV047160.1 | - | 1031 | 113 | |||
| JV652375.1 | - | 967 | 113 | |||
More Information
Set Membership: |
|

NEU1
Genomic Map