Assay Details
| Gene Symbol: SUOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113406.3 | XP_001113406.2 | 212 | 135 | ||
| XM_001113456.3 | XP_001113456.1 | 284 | 135 | |||
| XM_028829602.1 | XP_028685435.1 | 447 | 135 | |||
| GenBank mRNA | JU323720.1 | - | 124 | 135 | ||
| JV069632.1 | - | 197 | 135 | |||
More Information
Set Membership: |
|

SUOX
Genomic Map