Assay Details
| Gene Symbol: AK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094708.4 | XP_001094708.1 | 812 | 109 | ||
| XM_002800172.2 | XP_002800218.2 | 665 | 109 | |||
| XM_015116769.2 | XP_014972255.1 | 662 | 109 | |||
| GenBank mRNA | JU333325.1 | - | 670 | 109 | ||
| JV046410.1 | - | 532 | 109 | |||
| JV608478.1 | - | 661 | 109 | |||
More Information
Set Membership: |
|

AK1
Genomic Map