Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104583.1 | NP_001098053.1 | 808 | 111 | ||
| GenBank mRNA | EF057492.1 | - | 808 | 111 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SERPINC1
Genomic Map