Assay Details
| Gene Symbol: AVP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115061.4 | XP_001115061.2 | 915 | 81 | ||
| XM_028828268.1 | XP_028684101.1 | 801 | 81 | |||
| GenBank mRNA | JV688420.1 | - | 182 | 81 | ||
More Information
Set Membership: |
|

AVP
Genomic Map