Assay Details
| Gene Symbol: CYP21A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194627.2 | NP_001181556.2 | 1092 | 123 | ||
| XM_015135944.2 | XP_014991430.1 | 2670 | 123 | |||
More Information
Set Membership: |
|

CYP21A2
Genomic Map