Assay Details
| Gene Symbol: LDLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032906.1 | NP_001028078.1 | 195 | 116 | ||
| XM_015122900.2 | XP_014978386.2 | 231 | 116 | |||
| XM_028838328.1 | XP_028694161.1 | 853 | 116 | |||
| GenBank mRNA | AY466854.1 | - | 195 | 116 | ||
| JU471382.1 | - | 343 | 116 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LDLR
Genomic Map