Assay Details
| Gene Symbol: CD55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112168.3 | XP_001112168.1 | 966 | 148 | ||
| XM_015117701.2 | XP_014973187.1 | 966 | 148 | |||
| XM_015117708.1 | XP_014973194.1 | 966 | 148 | |||
| XM_015117716.1 | XP_014973202.1 | 966 | 148 | |||
| XM_015117723.2 | XP_014973209.1 | 1074 | 148 | |||
| XM_028852714.1 | XP_028708547.1 | 811 | 148 | |||
| XM_028852720.1 | XP_028708553.1 | 966 | 148 | |||
| GenBank mRNA | JU333166.1 | - | 871 | 148 | ||
| JU333167.1 | - | 871 | 148 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD55
Genomic Map