Assay Details
| Gene Symbol: ORM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100289.4 | XP_001100289.1 | 648 | 164 | ||
| XM_015117008.2 | XP_014972494.1 | 648 | 164 | |||
| XM_028834922.1 | XP_028690755.1 | 648 | 164 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ORM1
Genomic Map