Assay Details
| Gene Symbol: HTR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032966.1 | NP_001028138.1 | 613 | 96 | ||
| XM_015120877.2 | XP_014976363.2 | 1744 | 96 | |||
| XM_015120878.2 | XP_014976364.1 | 1016 | 96 | |||
| GenBank mRNA | JU322079.1 | - | 936 | 96 | ||
| S78209.1 | - | 613 | 96 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HTR2A
Genomic Map