Assay Details
| Gene Symbol: CD79B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261525.1 | NP_001248454.1 | 647 | 101 | ||
| XM_015120067.2 | XP_014975553.1 | 650 | 101 | |||
| XM_015120069.2 | XP_014975555.1 | 343 | 101 | |||
| XM_028835772.1 | XP_028691605.1 | 345 | 101 | |||
| GenBank mRNA | JU478000.1 | - | 647 | 101 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD79B
Genomic Map