Assay Details
| Gene Symbol: LOC100425407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844189.1 | XP_028700022.1 | 921 | 125 | ||
| GenBank mRNA | JU323020.1 | - | 907 | 125 | ||
| JU472825.1 | - | 893 | 125 | |||
| JV044383.1 | - | 853 | 125 | |||
| JV487714.1 | - | 89 | 125 | |||
| Gene Symbol: LOC698851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844191.1 | XP_028700024.1 | 1674 | 125 | ||
More Information
Set Membership: |
|

LOC100425407
Genomic Map