Assay Details
| Gene Symbol: CD247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077423.1 | NP_001070891.1 | 64 | 73 | ||
| XM_015124638.2 | XP_014980124.2 | 336 | 73 | |||
| GenBank mRNA | DQ437669.1 | - | 64 | 73 | ||
| DQ437670.1 | - | 64 | 73 | |||
| JU477807.1 | - | 208 | 73 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD247
Genomic Map