Assay Details
| Gene Symbol: ICAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195389.1 | NP_001182318.1 | 499 | 146 | ||
| XM_015120046.2 | XP_014975532.1 | 483 | 146 | |||
| GenBank mRNA | AF340054.1 | - | 282 | 146 | ||
| CB551343.1 | - | 515 | 146 | |||
| CN644791.1 | - | 141 | 146 | |||
| JU496308.1 | - | 1523 | 146 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ICAM2
Genomic Map