Assay Details
| Gene Symbol: PLA2G1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088684.4 | XP_001088684.1 | 300 | 134 | ||
| GenBank mRNA | JV653809.1 | - | 188 | 134 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLA2G1B
Genomic Map