Assay Details
| Gene Symbol: PPIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193973.2 | NP_001180902.1 | 293 | 143 | ||
| GenBank mRNA | JU326070.1 | - | 181 | 143 | ||
| JU476116.1 | - | 248 | 143 | |||
| JV045760.1 | - | 267 | 143 | |||
| JV717533.1 | - | 223 | 143 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPIC
Genomic Map