Assay Details
| Gene Symbol: PRRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261082.1 | NP_001248011.1 | 303 | 111 | ||
| XM_028841678.1 | XP_028697511.1 | 324 | 111 | |||
| XM_028841679.1 | XP_028697512.1 | 586 | 111 | |||
| GenBank mRNA | JU335492.1 | - | 303 | 111 | ||
More Information
Set Membership: |
|

PRRG1
Genomic Map