Assay Details
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115169.4 | XP_001115169.2 | 264 | 117 | ||
XM_015123428.1 | XP_014978914.1 | 339 | 117 | |||
XM_028840163.1 | XP_028695996.1 | 247 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015123428.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
