Assay Details
| Gene Symbol: COP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266265.1 | NP_001253194.1 | 1692 | 124 | ||
| XM_015123338.2 | XP_014978824.1 | 2025 | 124 | |||
| XM_015123346.2 | XP_014978832.1 | 1977 | 124 | |||
| XM_015123351.2 | XP_014978837.1 | 1965 | 124 | |||
| XM_015123363.2 | XP_014978849.1 | 1840 | 124 | |||
| XM_028843468.1 | XP_028699301.1 | 2091 | 124 | |||
| XM_028843469.1 | XP_028699302.1 | 2079 | 124 | |||
| XM_028843470.1 | XP_028699303.1 | 2031 | 124 | |||
| GenBank mRNA | JU327299.1 | - | 930 | 124 | ||
| JU476785.1 | - | 1632 | 124 | |||
| JU476786.1 | - | 1680 | 124 | |||
| JU476787.1 | - | 1692 | 124 | |||
| JV554568.1 | - | 1036 | 124 | |||
More Information
Set Membership: |
|

COP1
Genomic Map