Assay Details
| Gene Symbol: RAB11FIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089664.4 | XP_001089664.3 | 3669 | 148 | ||
| XM_015145164.2 | XP_015000650.1 | 1781 | 148 | |||
| GenBank mRNA | JU493968.1 | - | 960 | 148 | ||
| JV072091.1 | - | 352 | 148 | |||
More Information
Set Membership: |
|

RAB11FIP1
Genomic Map