Assay Details
| Gene Symbol: POLR1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103523.4 | XP_001103523.2 | 461 | 132 | ||
| GenBank mRNA | JU333411.1 | - | 407 | 132 | ||
| JU474697.1 | - | 414 | 132 | |||
| JV709578.1 | - | 415 | 132 | |||
More Information
Set Membership: |
|

POLR1F
Genomic Map