Assay Details
| Gene Symbol: LDLRAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837723.1 | XP_028693556.1 | 933 | 94 | ||
| XM_028837724.1 | XP_028693557.1 | 1026 | 94 | |||
| XM_028837725.1 | XP_028693558.1 | 996 | 94 | |||
| XM_028837726.1 | XP_028693559.1 | 813 | 94 | |||
| XM_028837727.1 | XP_028693560.1 | 743 | 94 | |||
| XM_028837728.1 | XP_028693561.1 | 873 | 94 | |||
| XM_028837731.1 | XP_028693564.1 | 1677 | 94 | |||
| XM_028837732.1 | XP_028693565.1 | 792 | 94 | |||
| XM_028837733.1 | XP_028693566.1 | 907 | 94 | |||
| GenBank mRNA | JU336266.1 | - | 1207 | 94 | ||
| JU471856.1 | - | 911 | 94 | |||
More Information
Set Membership: |
|

LDLRAD4
Genomic Map