Assay Details
| Gene Symbol: RABL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260670.2 | NP_001247599.1 | 287 | 81 | ||
| XM_015150634.2 | XP_015006120.2 | 650 | 81 | |||
| XM_015150635.2 | XP_015006121.2 | 760 | 81 | |||
| XM_015150638.2 | XP_015006124.2 | 404 | 81 | |||
| XM_028827398.1 | XP_028683231.1 | 908 | 81 | |||
| XM_028827399.1 | XP_028683232.1 | 904 | 81 | |||
| XM_028827400.1 | XP_028683233.1 | 622 | 81 | |||
| XM_028827401.1 | XP_028683234.1 | 513 | 81 | |||
| XM_028827402.1 | XP_028683235.1 | 798 | 81 | |||
| XM_028827403.1 | XP_028683236.1 | 644 | 81 | |||
| XM_028827404.1 | XP_028683237.1 | 185 | 81 | |||
| XM_028827406.1 | XP_028683239.1 | 653 | 81 | |||
| XM_028827407.1 | XP_028683240.1 | 652 | 81 | |||
| XM_028827408.1 | XP_028683241.1 | 405 | 81 | |||
| XM_028827409.1 | XP_028683242.1 | 656 | 81 | |||
| GenBank mRNA | JU323199.1 | - | 370 | 81 | ||
| JU471428.1 | - | 295 | 81 | |||
| JV046060.1 | - | 287 | 81 | |||
| JV567305.1 | - | 433 | 81 | |||
| JV704339.1 | - | 514 | 81 | |||
| Gene Symbol: LOC114677631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847684.1 | XP_028703517.1 | 323 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028827406.1, XM_028827407.1, XM_028827408.1, XM_028827409.1 transcripts.
Set Membership: |
|

RABL2B
Genomic Map