Assay Details
| Gene Symbol: RABL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260670.2 | NP_001247599.1 | 746 | 122 | ||
| XM_015150634.2 | XP_015006120.2 | 1109 | 119 | |||
| XM_015150635.2 | XP_015006121.2 | 1219 | 119 | |||
| XM_015150638.2 | XP_015006124.2 | 860 | 119 | |||
| XM_028827398.1 | XP_028683231.1 | 1367 | 119 | |||
| XM_028827399.1 | XP_028683232.1 | 1363 | 119 | |||
| XM_028827400.1 | XP_028683233.1 | 1081 | 119 | |||
| XM_028827401.1 | XP_028683234.1 | 972 | 119 | |||
| XM_028827402.1 | XP_028683235.1 | 1257 | 119 | |||
| XM_028827403.1 | XP_028683236.1 | 1103 | 119 | |||
| XM_028827405.1 | XP_028683238.1 | 1257 | 119 | |||
| XM_028827406.1 | XP_028683239.1 | 1149 | 119 | |||
| XM_028827407.1 | XP_028683240.1 | 1031 | 119 | |||
| XM_028827408.1 | XP_028683241.1 | 784 | 119 | |||
| XM_028827409.1 | XP_028683242.1 | 1032 | 119 | |||
| GenBank mRNA | DQ148034.1 | - | 244 | 122 | ||
| JU323199.1 | - | 829 | 122 | |||
| JU471428.1 | - | 754 | 122 | |||
| JV046060.1 | - | 746 | 122 | |||
| JV567305.1 | - | 892 | 122 | |||
| JV704339.1 | - | 973 | 122 | |||
| Gene Symbol: LOC114677631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847684.1 | XP_028703517.1 | 782 | 122 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RABL2B
Genomic Map