Assay Details
Gene Symbol: BICD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257956.1 | NP_001244885.1 | 2311 | 86 | ||
XM_015117247.2 | XP_014972733.1 | 2375 | 86 | |||
GenBank mRNA | JU321384.1 | - | 2311 | 86 | ||
JU333631.1 | - | 2311 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
