Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092529.4 | XP_001092529.2 | 1064 | 144 | ||
| GenBank mRNA | JU325749.1 | - | 395 | 144 | ||
| JV715457.1 | - | 246 | 144 | |||
More Information
Set Membership: |
|

DBX2
Genomic Map