Assay Details
| Gene Symbol: GPNMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266650.1 | NP_001253579.1 | 618 | 81 | ||
| XM_015133861.2 | XP_014989347.1 | 771 | 81 | |||
| XM_015133862.2 | XP_014989348.1 | 860 | 81 | |||
| XM_015133863.2 | XP_014989349.1 | 772 | 81 | |||
| XM_015133864.2 | XP_014989350.1 | 561 | 81 | |||
| XM_015133865.2 | XP_014989351.1 | 840 | 81 | |||
| GenBank mRNA | JU332515.1 | - | 638 | 81 | ||
| JU473990.1 | - | 618 | 81 | |||
More Information
Set Membership: |
|

GPNMB
Genomic Map