Assay Details
| Gene Symbol: PLAUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124159.2 | XP_014979645.1 | 1124 | 79 | ||
| XM_028839257.1 | XP_028695090.1 | 2216 | 79 | |||
| GenBank mRNA | JU488228.1 | - | 412 | 79 | ||
| JV047392.1 | - | 578 | 79 | |||
| JV635727.1 | - | 652 | 79 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLAUR
Genomic Map