Assay Details
| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260577.1 | NP_001247506.1 | 911 | 64 | ||
| GenBank mRNA | JU322096.1 | - | 911 | 64 | ||
| JV059946.1 | - | 185 | 64 | |||
More Information
Set Membership: |
|

PLCXD3
Genomic Map