Assay Details
| Gene Symbol: HMCES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194040.1 | NP_001180969.1 | 693 | 92 | ||
| XM_001095958.4 | XP_001095958.1 | 650 | 92 | |||
| XM_015130134.2 | XP_014985620.1 | 611 | 92 | |||
| XM_028843211.1 | XP_028699044.1 | 781 | 92 | |||
| XM_028843212.1 | XP_028699045.1 | 603 | 92 | |||
| GenBank mRNA | JU335538.1 | - | 520 | 92 | ||
| JU476466.1 | - | 523 | 92 | |||
| JV513579.1 | - | 477 | 92 | |||
| JV587837.1 | - | 519 | 92 | |||
| JV656419.1 | - | 424 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HMCES
Genomic Map