Assay Details
| Gene Symbol: IP6K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257763.1 | NP_001244692.1 | 744 | 61 | ||
| XM_015130794.2 | XP_014986280.1 | 1138 | 61 | |||
| XM_015130795.2 | XP_014986281.1 | 579 | 61 | |||
| XM_015130796.2 | XP_014986282.1 | 582 | 61 | |||
| XM_028843239.1 | XP_028699072.1 | 578 | 61 | |||
| GenBank mRNA | JU337334.1 | - | 744 | 61 | ||
| JU478036.1 | - | 740 | 61 | |||
| JV043613.1 | - | 750 | 61 | |||
| JV666759.1 | - | 752 | 61 | |||
More Information
Set Membership: |
|

IP6K1
Genomic Map