Assay Details
| Gene Symbol: SDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028831950.1 | XP_028687783.1 | 444 | 121 | ||
| GenBank mRNA | JV043488.1 | - | 444 | 121 | ||
| JV636312.1 | - | 397 | 121 | |||
More Information
Set Membership: |
|

SDC1
Genomic Map