Assay Details
| Gene Symbol: C18H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084520.4 | XP_001084520.1 | 112 | 150 | ||
| XM_001084644.4 | XP_001084644.1 | 299 | 150 | |||
| GenBank mRNA | JU320430.1 | - | 229 | 150 | ||
| JU471827.1 | - | 280 | 150 | |||
| JV043308.1 | - | 246 | 150 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001084520.4, XM_001084644.4 transcripts.
Set Membership: |
|

C18H18orf25
Genomic Map