Assay Details
| Gene Symbol: ISX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194582.2 | NP_001181511.2 | 556 | 126 | ||
| XM_028827423.1 | XP_028683256.1 | 1160 | 126 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ISX| Gene Symbol: ISX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194582.2 | NP_001181511.2 | 556 | 126 | ||
| XM_028827423.1 | XP_028683256.1 | 1160 | 126 | |||
Set Membership: |
|