Assay Details
Gene Symbol: RAB19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261554.1 | NP_001248483.1 | 485 | 114 | ||
GenBank mRNA | JU475583.1 | - | 485 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
