Assay Details
| Gene Symbol: FAM168B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194008.2 | NP_001180937.1 | 388 | 94 | ||
| GenBank mRNA | JU320484.1 | - | 381 | 94 | ||
| JU471185.1 | - | 390 | 94 | |||
| JV043682.1 | - | 388 | 94 | |||
More Information
Set Membership: |
|

FAM168B
Genomic Map