Assay Details
| Gene Symbol: TSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194867.2 | NP_001181796.2 | 429 | 108 | ||
| XM_001117184.4 | XP_001117184.2 | 594 | 108 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPO2| Gene Symbol: TSPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194867.2 | NP_001181796.2 | 429 | 108 | ||
| XM_001117184.4 | XP_001117184.2 | 594 | 108 | |||
Set Membership: |
|