Assay Details
| Gene Symbol: LIPJ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147641.2 | XP_015003127.1 | 2481 | 133 | ||
| XM_015147642.2 | XP_015003128.1 | 2481 | 133 | |||
More Information
Set Membership: |
|

LIPJ
Genomic Map