Assay Details
| Gene Symbol: C1H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111453.4 | XP_001111453.1 | 327 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001111453.4 transcripts.
Set Membership: |
|

C1H1orf189
Genomic Map