Assay Details
| Gene Symbol: AFMID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105238.4 | XP_001105238.4 | 805 | 73 | ||
| XM_028836859.1 | XP_028692692.1 | 810 | 73 | |||
| XM_028836860.1 | XP_028692693.1 | 710 | 73 | |||
| XM_028836862.1 | XP_028692695.1 | 705 | 73 | |||
| GenBank mRNA | JU338677.1 | - | 416 | 73 | ||
More Information
Set Membership: |
|

AFMID
Genomic Map