Assay Details
| Gene Symbol: MYBPHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089938.4 | XP_001089938.2 | 308 | 92 | ||
| XM_015147705.2 | XP_015003191.2 | 308 | 92 | |||
| XM_015147715.2 | XP_015003201.2 | 308 | 92 | |||
| XM_028830677.1 | XP_028686510.1 | 308 | 92 | |||
| GenBank mRNA | JV059907.1 | - | 227 | 92 | ||
More Information
Set Membership: |
|

MYBPHL
Genomic Map