Assay Details
| Gene Symbol: CNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121146.2 | XP_014976632.1 | 621 | 98 | ||
| XM_015121147.2 | XP_014976633.1 | 625 | 98 | |||
| GenBank mRNA | JU322860.1 | - | 411 | 98 | ||
More Information
Set Membership: |
|

CNMD
Genomic Map